The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277115.1 | NP_001264044.1 | 7 - 8 | 1459 | 104 |
GenBank mRNA | AJ320497.1 | - | 7 - 8 | 1459 | 104 |