The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270476.1 | NP_001257405.1 | 8 - 9 | 1117 | 96 |
NM_006245.3 | NP_006236.1 | 8 - 9 | 1100 | 96 | |
NM_180976.2 | NP_851307.1 | 8 - 9 | 1004 | 96 | |
NM_180977.2 | NP_851308.1 | 6 - 7 | 782 | 96 | |
GenBank mRNA | AB000634.1 | - | 8 - 9 | 935 | 96 |
AB000635.1 | - | 6 - 7 | 629 | 96 | |
AB209861.1 | - | 6 - 7 | 679 | 96 | |
AB451342.1 | - | 8 - 9 | 921 | 96 | |
AB451357.1 | - | 8 - 9 | 921 | 96 | |
AK122701.1 | - | 8 - 9 | 994 | 96 | |
AK290604.1 | - | 8 - 9 | 913 | 96 | |
AK291797.1 | - | 8 - 9 | 1008 | 96 | |
AK299696.1 | - | 8 - 9 | 973 | 96 | |
BC001095.2 | - | 8 - 9 | 984 | 96 | |
BC001175.1 | - | 8 - 9 | 911 | 96 | |
BC010692.1 | - | 8 - 9 | 994 | 96 | |
D78360.1 | - | 8 - 9 | 902 | 96 | |
L76702.1 | - | 8 - 9 | 1108 | 96 |