The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017011715.1 | XP_016867204.1 | 3 - 4 | 501 | 110 |
XM_017011716.1 | XP_016867205.1 | 3 - 4 | 1244 | 110 | |
XM_017011717.1 | XP_016867206.1 | 2 - 3 | 227 | 110 | |
GenBank mRNA | AK298183.1 | - | 3 - 4 | 437 | 110 |