The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100623.1 | NP_001094093.1 | 6 - 7 | 984 | 112 |
NM_001267062.1 | NP_001253991.1 | 6 - 7 | 984 | 112 | |
NM_001309448.1 | NP_001296377.1 | 5 - 6 | 939 | 112 | |
NM_001309450.1 | NP_001296379.1 | 4 - 5 | 841 | 112 | |
NM_001309452.1 | NP_001296381.1 | 7 - 8 | 1082 | 112 | |
XM_017004409.1 | XP_016859898.1 | 5 - 6 | 452 | 112 | |
XM_017004412.1 | XP_016859901.1 | 7 - 8 | 1588 | 112 | |
GenBank mRNA | AK093730.1 | - | 6 - 7 | 1219 | 112 |
AK123710.1 | - | 6 - 7 | 941 | 112 | |
AK296008.1 | - | 5 - 6 | 448 | 112 | |
BC013991.2 | - | 6 - 7 | 477 | 112 | |
BC050695.1 | - | 6 - 7 | 479 | 112 | |
BQ052811.1 | - | 6 - 7 | 476 | 112 | |
BU174451.1 | - | 7 - 8 | 563 | 112 | |
CK000843.1 | - | 4 - 5 | 344 | 112 |