Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFHC2
Entrez Gene ID
80258
Gene Name
EF-hand domain containing 2
Gene Aliases
MRX74, dJ1158H2.1
Chromosome Location
Chr.X: 44146254 - 44343677 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025184.3 NP_079460.2 5 - 6 945 83
XM_006724562.2 XP_006724625.1 4 - 5 658 83
GenBank mRNA AK026496.1 - 5 - 6 798 83
AK291187.1 - 5 - 6 1086 83
AK292967.1 - 5 - 6 929 83
AK297714.1 - 5 - 6 668 83
BC031039.1 - 5 - 6 940 83
CR749363.1 - 1 - 2 214 83

View Full Product Details