Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFHC2
Entrez Gene ID
80258
Gene Name
EF-hand domain containing 2
Gene Aliases
MRX74, dJ1158H2.1
Chromosome Location
Chr.X: 44146254 - 44343677 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025184.3 NP_079460.2 7 - 8 1203 132
XM_006724562.2 XP_006724625.1 6 - 7 916 132
GenBank mRNA AK026496.1 - 7 - 8 1056 132
AK291187.1 - 7 - 8 1344 132
AK292967.1 - 7 - 8 1187 132
BC031039.1 - 7 - 8 1198 132
CR749363.1 - 2 - 3 864 132

View Full Product Details