The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308025.1 | NP_001294954.1 | 19 - 20 | 3304 | 112 |
NM_152259.3 | NP_689472.3 | 19 - 20 | 3307 | 112 | |
GenBank mRNA | AK123612.1 | - | 12 - 13 | 1407 | 112 |
BX538335.1 | - | 6 - 7 | 608 | 112 | |
GQ227787.1 | - | 19 - 20 | 3304 | 112 |