The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014809.2 | NP_001014809.1 | 7 - 8 | 1147 | 91 |
NM_001288661.1 | NP_001275590.1 | 7 - 8 | 1254 | 91 | |
NM_001288662.1 | NP_001275591.1 | 7 - 8 | 1077 | 91 | |
NM_001313.4 | NP_001304.1 | 7 - 8 | 1041 | 91 | |
GenBank mRNA | AK094725.1 | - | 7 - 8 | 806 | 91 |
AK122754.1 | - | 7 - 8 | 1077 | 91 | |
AK127754.1 | - | 8 - 9 | 793 | 91 | |
BC000252.1 | - | 7 - 8 | 906 | 91 | |
BC007613.1 | - | 7 - 8 | 906 | 91 | |
BC007898.2 | - | 3 - 4 | 189 | 91 | |
BT006806.1 | - | 7 - 8 | 691 | 91 | |
D78012.1 | - | 7 - 8 | 841 | 91 | |
DQ206871.1 | - | 7 - 8 | 1033 | 91 | |
KJ534816.1 | - | 7 - 8 | 1033 | 91 | |
KJ534817.1 | - | 8 - 9 | 940 | 91 | |
U17278.1 | - | 5 - 6 | 518 | 91 |