Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLGN4Y
Entrez Gene ID
22829
Gene Name
neuroligin 4, Y-linked
Gene Aliases
-
Chromosome Location
Chr.Y: 14522578 - 14844945 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164238.1 NP_001157710.1 1 - 2 570 149
NR_046355.1 2 - 3 650 149
XM_006724874.2 XP_006724937.1 2 - 3 895 149
XM_011531424.2 XP_011529726.1 2 - 3 1213 149
XM_011531425.2 XP_011529727.1 2 - 3 985 149
XM_011531426.2 XP_011529728.1 2 - 3 946 149
XM_011531427.1 XP_011529729.1 2 - 3 700 149
XM_011531428.1 XP_011529730.1 3 - 4 881 149
XM_011531429.2 XP_011529731.1 2 - 3 1496 149
XM_011531430.2 XP_011529732.1 2 - 3 834 149
XM_017030041.1 XP_016885530.1 2 - 3 984 149
GenBank mRNA AK295117.1 - 2 - 3 650 149
BC032567.1 - 1 - 2 373 149

View Full Product Details