The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143981.1 | NP_001137453.1 | 10 - 11 | 1504 | 88 |
NM_001143982.1 | NP_001137454.1 | 10 - 11 | 1501 | 88 | |
NM_001143983.2 | NP_001137455.2 | 8 - 9 | 1264 | 88 | |
NM_145234.3 | NP_660277.2 | 10 - 11 | 1346 | 88 | |
XM_005262221.1 | XP_005262278.1 | 10 - 11 | 1321 | 88 | |
XM_005262222.3 | XP_005262279.1 | 10 - 11 | 1318 | 88 | |
XM_005262223.1 | XP_005262280.1 | 10 - 11 | 1283 | 88 | |
XM_005262224.1 | XP_005262281.1 | 10 - 11 | 1302 | 88 | |
XM_017029959.1 | XP_016885448.1 | 10 - 11 | 1306 | 88 | |
GenBank mRNA | AB209548.1 | - | 9 - 10 | 1094 | 88 |
AK092245.1 | - | 7 - 8 | 931 | 88 | |
AK293106.1 | - | 10 - 11 | 1251 | 88 | |
AK297563.1 | - | 8 - 9 | 1025 | 88 | |
AK312270.1 | - | 10 - 11 | 1262 | 88 | |
AY608914.1 | - | 9 - 10 | 1178 | 88 | |
BC002909.2 | - | 10 - 11 | 1263 | 88 |