The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015520.1 | NP_056335.1 | 24 - 25 | 4339 | 92 |
XM_005265574.3 | XP_005265631.1 | 24 - 25 | 4344 | 92 | |
XM_006713414.3 | XP_006713477.1 | 24 - 25 | 4350 | 92 | |
XM_017007502.1 | XP_016862991.1 | 24 - 25 | 4347 | 92 | |
XM_017007503.1 | XP_016862992.1 | 24 - 25 | 4347 | 92 | |
XM_017007504.1 | XP_016862993.1 | 24 - 25 | 4344 | 92 | |
XM_017007505.1 | XP_016862994.1 | 24 - 25 | 4344 | 92 | |
XM_017007506.1 | XP_016862995.1 | 24 - 25 | 4308 | 92 | |
GenBank mRNA | AF401654.1 | - | 24 - 25 | 3813 | 92 |