Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF20L1
Entrez Gene ID
51105
Gene Name
PHD finger protein 20-like 1
Gene Aliases
CGI-72, TDRD20B, URLC1
Chromosome Location
Chr.8: 132775358 - 132848807 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277196.1 NP_001264125.1 7 - 8 1095 67
NM_016018.4 NP_057102.4 8 - 9 1173 67
NM_198513.1 NP_940915.1 7 - 8 1091 67
XM_005250931.1 XP_005250988.1 8 - 9 1158 67
XM_011517075.1 XP_011515377.1 8 - 9 1158 67
XM_011517076.1 XP_011515378.1 8 - 9 1154 67
XM_011517077.1 XP_011515379.1 8 - 9 1155 67
XM_011517078.1 XP_011515380.1 8 - 9 1146 67
XM_011517079.1 XP_011515381.1 8 - 9 1143 67
XM_011517080.1 XP_011515382.1 7 - 8 1068 67
XM_011517081.1 XP_011515383.1 7 - 8 1065 67
XM_011517088.1 XP_011515390.1 8 - 9 1158 67
XM_017013509.1 XP_016868998.1 8 - 9 1139 67
XM_017013510.1 XP_016868999.1 7 - 8 1064 67
XM_017013511.1 XP_016869000.1 7 - 8 1061 67
XM_017013512.1 XP_016869001.1 7 - 8 1061 67
XM_017013513.1 XP_016869002.1 8 - 9 3981 67
XM_017013514.1 XP_016869003.1 8 - 9 3977 67
XM_017013516.1 XP_016869005.1 8 - 9 1143 67
XM_017013517.1 XP_016869006.1 7 - 8 1065 67
GenBank mRNA AB101203.1 - 8 - 9 1140 67
AF151830.1 - 5 - 6 527 67
AK023711.1 - 2 - 3 618 67
AK290506.1 - 7 - 8 1065 67
BC002822.2 - 8 - 9 1131 67
BC015211.1 - 7 - 8 1041 67

View Full Product Details