The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144703.2 | NP_653304.2 | 5 - 6 | 880 | 66 |
XM_005260302.3 | XP_005260359.1 | 6 - 7 | 992 | 66 | |
XM_011528605.2 | XP_011526907.1 | 7 - 8 | 1184 | 66 | |
XM_011528606.2 | XP_011526908.1 | 7 - 8 | 1102 | 66 | |
XM_011528608.2 | XP_011526910.1 | 6 - 7 | 1073 | 66 | |
XM_011528609.2 | XP_011526911.1 | 5 - 6 | 954 | 66 | |
XM_011528612.2 | XP_011526914.1 | 7 - 8 | 854 | 66 | |
XM_011528613.2 | XP_011526915.1 | 6 - 7 | 714 | 66 | |
XM_011528614.2 | XP_011526916.1 | 6 - 7 | 927 | 66 | |
XM_011528615.2 | XP_011526917.1 | 3 - 4 | 411 | 66 | |
XM_017027688.1 | XP_016883177.1 | 4 - 5 | 787 | 66 | |
XM_017027689.1 | XP_016883178.1 | 4 - 5 | 706 | 66 | |
XM_017027690.1 | XP_016883179.1 | 4 - 5 | 528 | 66 | |
GenBank mRNA | AK058202.1 | - | 7 - 8 | 859 | 66 |
BC035702.1 | - | 5 - 6 | 945 | 66 |