Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRDM8
Entrez Gene ID
56978
Gene Name
PR domain 8
Gene Aliases
EPM10, PFM5
Chromosome Location
Chr.4: 80183418 - 80204329 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005263145.2 XP_005263202.1 1 - 2 169 135

View Full Product Details