Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHB2
Entrez Gene ID
55041
Gene Name
pleckstrin homology domain containing B2
Gene Aliases
EVT2
Chromosome Location
Chr.2: 131104847 - 131149852 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267063.1 NP_001253992.1 6 - 7 987 109
NM_001309451.1 NP_001296380.1 5 - 6 942 109
NM_017958.2 NP_060428.2 6 - 7 987 109
GenBank mRNA AK000790.1 - 6 - 7 498 109
AK223338.1 - 6 - 7 498 109
BP362174.1 - 5 - 6 453 109

View Full Product Details