The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015245.2 | NP_056060.2 | 22 - 23 | 3451 | 57 |
XM_005248964.3 | XP_005249021.1 | 23 - 24 | 3502 | 57 | |
XM_006715036.2 | XP_006715099.1 | 23 - 24 | 3505 | 57 | |
XM_011514431.2 | XP_011512733.1 | 23 - 24 | 3505 | 57 | |
XM_011514432.2 | XP_011512734.1 | 23 - 24 | 3502 | 57 | |
XM_011514433.1 | XP_011512735.1 | 23 - 24 | 3505 | 57 | |
XM_011514434.2 | XP_011512736.1 | 22 - 23 | 3442 | 57 | |
XM_011514435.2 | XP_011512737.1 | 22 - 23 | 3439 | 57 | |
XM_011514436.2 | XP_011512738.1 | 23 - 24 | 7074 | 57 | |
XM_011514437.2 | XP_011512739.1 | 13 - 14 | 3330 | 57 | |
XM_017010593.1 | XP_016866082.1 | 13 - 14 | 1928 | 57 | |
GenBank mRNA | BC049213.1 | - | 11 - 12 | 1294 | 57 |
BC132832.1 | - | 22 - 23 | 3388 | 57 | |
BX094969.1 | - | 5 - 6 | 580 | 57 | |
D86982.1 | - | 22 - 23 | 3448 | 57 |