The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001254718.1 | NP_001241647.1 | 23 - 24 | 2731 | 78 |
NM_001254719.1 | NP_001241648.1 | 22 - 23 | 2677 | 78 | |
NM_001254720.1 | NP_001241649.1 | 21 - 22 | 2641 | 78 | |
NM_001254721.1 | NP_001241650.1 | 21 - 22 | 2620 | 78 | |
NM_001254722.1 | NP_001241651.1 | 20 - 21 | 2599 | 78 | |
NM_001254723.1 | NP_001241652.1 | 21 - 22 | 2638 | 78 | |
NM_002465.3 | NP_002456.2 | 24 - 25 | 2752 | 78 | |
NM_206819.2 | NP_996555.1 | 24 - 25 | 2752 | 78 | |
NM_206820.2 | NP_996556.1 | 23 - 24 | 2731 | 78 | |
NM_206821.2 | NP_996557.1 | 22 - 23 | 2677 | 78 | |
XM_005268876.4 | XP_005268933.1 | 23 - 24 | 2686 | 78 | |
XM_006719405.3 | XP_006719468.1 | 25 - 26 | 2667 | 78 | |
XM_006719406.3 | XP_006719469.1 | 25 - 26 | 2667 | 78 | |
XM_006719407.3 | XP_006719470.1 | 24 - 25 | 2740 | 78 | |
XM_006719408.3 | XP_006719471.1 | 24 - 25 | 2613 | 78 | |
XM_006719409.3 | XP_006719472.1 | 23 - 24 | 2693 | 78 | |
XM_006719410.3 | XP_006719473.1 | 25 - 26 | 2667 | 78 | |
XM_006719411.3 | XP_006719474.1 | 23 - 24 | 2693 | 78 | |
XM_017019315.1 | XP_016874804.1 | 24 - 25 | 2610 | 78 | |
XM_017019316.1 | XP_016874805.1 | 25 - 26 | 2667 | 78 | |
XM_017019317.1 | XP_016874806.1 | 22 - 23 | 2639 | 78 | |
XM_017019318.1 | XP_016874807.1 | 23 - 24 | 2556 | 78 | |
XM_017019319.1 | XP_016874808.1 | 22 - 23 | 2636 | 78 | |
XM_017019320.1 | XP_016874809.1 | 21 - 22 | 2735 | 78 | |
XM_017019321.1 | XP_016874810.1 | 21 - 22 | 2582 | 78 | |
XM_017019322.1 | XP_016874811.1 | 19 - 20 | 2624 | 78 | |
GenBank mRNA | AK057284.1 | - | 4 - 5 | 796 | 78 |
AK292976.1 | - | 24 - 25 | 2712 | 78 | |
AK295727.1 | - | 20 - 21 | 2559 | 78 | |
AK296681.1 | - | 23 - 24 | 2695 | 78 | |
AK303401.1 | - | 21 - 22 | 2598 | 78 | |
AL831993.2 | - | 24 - 25 | 2687 | 78 | |
AL832000.2 | - | 24 - 25 | 2687 | 78 | |
AL832332.2 | - | 24 - 25 | 2687 | 78 | |
AL832350.2 | - | 24 - 25 | 2687 | 78 | |
BC092418.1 | - | 22 - 23 | 2624 | 78 | |
BC117217.1 | - | 20 - 21 | 2534 | 78 | |
BC143495.1 | - | 21 - 22 | 2555 | 78 | |
BC143502.1 | - | 20 - 21 | 2534 | 78 | |
BC143503.1 | - | 22 - 23 | 2612 | 78 | |
BC143504.1 | - | 21 - 22 | 2576 | 78 | |
BX649073.2 | - | 25 - 26 | 2782 | 78 | |
X73114.1 | - | 22 - 23 | 2618 | 78 |