Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYBPC1
Entrez Gene ID
4604
Gene Name
myosin binding protein C, slow type
Gene Aliases
LCCS4, MYBPCC, MYBPCS
Chromosome Location
Chr.12: 101594849 - 101695841 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001254718.1 NP_001241647.1 24 - 25 2921 80
NM_001254719.1 NP_001241648.1 23 - 24 2867 80
NM_001254720.1 NP_001241649.1 22 - 23 2831 80
NM_001254721.1 NP_001241650.1 22 - 23 2810 80
NM_001254722.1 NP_001241651.1 21 - 22 2789 80
NM_001254723.1 NP_001241652.1 22 - 23 2828 80
NM_002465.3 NP_002456.2 25 - 26 2942 80
NM_206819.2 NP_996555.1 25 - 26 2942 80
NM_206820.2 NP_996556.1 24 - 25 2921 80
NM_206821.2 NP_996557.1 23 - 24 2867 80
XM_005268876.4 XP_005268933.1 24 - 25 2876 80
XM_006719405.3 XP_006719468.1 26 - 27 2857 80
XM_006719406.3 XP_006719469.1 26 - 27 2857 80
XM_006719407.3 XP_006719470.1 25 - 26 2930 80
XM_006719408.3 XP_006719471.1 25 - 26 2803 80
XM_006719409.3 XP_006719472.1 24 - 25 2883 80
XM_006719410.3 XP_006719473.1 26 - 27 2857 80
XM_006719411.3 XP_006719474.1 24 - 25 2883 80
XM_017019315.1 XP_016874804.1 25 - 26 2800 80
XM_017019316.1 XP_016874805.1 26 - 27 2857 80
XM_017019317.1 XP_016874806.1 23 - 24 2829 80
XM_017019318.1 XP_016874807.1 24 - 25 2746 80
XM_017019319.1 XP_016874808.1 23 - 24 2826 80
XM_017019320.1 XP_016874809.1 22 - 23 2925 80
XM_017019321.1 XP_016874810.1 22 - 23 2772 80
XM_017019322.1 XP_016874811.1 20 - 21 2814 80
GenBank mRNA AK057284.1 - 5 - 6 986 80
AK292976.1 - 25 - 26 2902 80
AK295727.1 - 21 - 22 2749 80
AK296681.1 - 24 - 25 2885 80
AL831993.2 - 25 - 26 2877 80
AL832000.2 - 25 - 26 2877 80
AL832350.2 - 25 - 26 2877 80
AL832354.2 - 25 - 26 2880 80
AL832456.2 - 25 - 26 2877 80
AL834249.2 - 25 - 26 2877 80
BC092418.1 - 23 - 24 2814 80
BC117217.1 - 21 - 22 2724 80
BC143495.1 - 22 - 23 2745 80
BC143502.1 - 21 - 22 2724 80
BC143503.1 - 23 - 24 2802 80
BC143504.1 - 22 - 23 2766 80
BX649073.2 - 26 - 27 2972 80
X73114.1 - 23 - 24 2808 80

View Full Product Details