Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYBPC1
Entrez Gene ID
4604
Gene Name
myosin binding protein C, slow type
Gene Aliases
LCCS4, MYBPCC, MYBPCS
Chromosome Location
Chr.12: 101594849 - 101695841 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001254718.1 NP_001241647.1 7 - 8 675 83
NM_001254719.1 NP_001241648.1 7 - 8 675 83
NM_001254720.1 NP_001241649.1 6 - 7 639 83
NM_001254722.1 NP_001241651.1 5 - 6 597 83
NM_001254723.1 NP_001241652.1 6 - 7 636 83
NM_002465.3 NP_002456.2 9 - 10 750 83
NM_206819.2 NP_996555.1 9 - 10 750 83
NM_206820.2 NP_996556.1 7 - 8 675 83
NM_206821.2 NP_996557.1 7 - 8 675 83
XM_005268876.4 XP_005268933.1 8 - 9 684 83
XM_006719405.3 XP_006719468.1 9 - 10 611 83
XM_006719406.3 XP_006719469.1 9 - 10 611 83
XM_006719407.3 XP_006719470.1 8 - 9 684 83
XM_006719408.3 XP_006719471.1 9 - 10 611 83
XM_006719409.3 XP_006719472.1 7 - 8 637 83
XM_006719410.3 XP_006719473.1 9 - 10 611 83
XM_006719411.3 XP_006719474.1 7 - 8 637 83
XM_017019316.1 XP_016874805.1 9 - 10 611 83
XM_017019317.1 XP_016874806.1 7 - 8 637 83
XM_017019320.1 XP_016874809.1 5 - 6 679 83
GenBank mRNA AK056587.1 - 8 - 9 674 83
AK292976.1 - 9 - 10 710 83
AK295727.1 - 5 - 6 557 83
AK296681.1 - 7 - 8 639 83
AK303401.1 - 6 - 7 596 83
AL831993.2 - 9 - 10 685 83
AL832000.2 - 9 - 10 685 83
AL832332.2 - 9 - 10 685 83
AL832350.2 - 9 - 10 685 83
AL832354.2 - 9 - 10 687 83
AL832456.2 - 9 - 10 685 83
AL834249.2 - 9 - 10 685 83
BC015187.1 - 9 - 10 710 83
BC065720.1 - 5 - 6 560 83
BC092418.1 - 7 - 8 622 83
BC117217.1 - 5 - 6 532 83
BC143502.1 - 5 - 6 532 83
BC143503.1 - 7 - 8 610 83
BC143504.1 - 6 - 7 574 83
BX649073.2 - 10 - 11 781 83
CD614901.1 - 4 - 5 375 83
DA433791.1 - 7 - 8 639 83
X66276.1 - 7 - 8 628 83
X73114.1 - 7 - 8 616 83

View Full Product Details