The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304743.1 | NP_001291672.1 | 6 - 7 | 1344 | 95 |
NM_014720.3 | NP_055535.2 | 6 - 7 | 1344 | 95 | |
XM_011540401.2 | XP_011538703.1 | 6 - 7 | 1356 | 95 | |
GenBank mRNA | AB002804.1 | - | 6 - 7 | 1015 | 95 |
AF273048.1 | - | 6 - 7 | 817 | 95 | |
AK302855.1 | - | 6 - 7 | 777 | 95 | |
AK307389.1 | - | 7 - 8 | 896 | 95 | |
BC047762.1 | - | 6 - 7 | 1337 | 95 | |
BC047885.1 | - | 6 - 7 | 1344 | 95 | |
BC064804.1 | - | 6 - 7 | 1340 | 95 | |
BC111565.1 | - | 6 - 7 | 814 | 95 | |
D86959.1 | - | 6 - 7 | 1294 | 95 |