The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018074.4 | NP_060544.2 | 2 - 3 | 165 | 117 |
GenBank mRNA | AF091080.1 | - | 2 - 3 | 153 | 117 |
AK001236.1 | - | 2 - 3 | 189 | 117 | |
AK023988.1 | - | 2 - 3 | 165 | 117 | |
BC000561.2 | - | 2 - 3 | 172 | 117 | |
BC019096.2 | - | 2 - 3 | 210 | 117 |