Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC94
Entrez Gene ID
55702
Gene Name
coiled-coil domain containing 94
Gene Aliases
-
Chromosome Location
Chr.19: 4247114 - 4269088 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018074.4 NP_060544.2 5 - 6 623 121
GenBank mRNA AF091080.1 - 5 - 6 611 121
AK001236.1 - 5 - 6 647 121
AK023988.1 - 5 - 6 623 121
BC000561.2 - 5 - 6 630 121
BC019096.2 - 5 - 6 668 121

View Full Product Details