The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111038.1 | NP_001104508.1 | 6 - 7 | 739 | 147 |
NM_001287150.1 | NP_001274079.1 | 6 - 7 | 739 | 147 | |
NM_145032.3 | NP_659469.3 | 6 - 7 | 916 | 147 | |
NR_105043.1 | 7 - 8 | 830 | 147 | ||
XM_005250205.3 | XP_005250262.1 | 7 - 8 | 822 | 147 | |
XM_005250207.3 | XP_005250264.1 | 7 - 8 | 822 | 147 | |
XM_005250208.3 | XP_005250265.1 | 7 - 8 | 822 | 147 | |
XM_005250209.2 | XP_005250266.1 | 6 - 7 | 720 | 147 | |
XM_011515928.2 | XP_011514230.1 | 7 - 8 | 822 | 147 | |
XM_011515929.2 | XP_011514231.1 | 7 - 8 | 822 | 147 | |
XM_011515930.2 | XP_011514232.1 | 6 - 7 | 786 | 147 | |
XM_011515932.2 | XP_011514234.1 | 7 - 8 | 822 | 147 | |
XM_017011850.1 | XP_016867339.1 | 7 - 8 | 766 | 147 | |
XM_017011851.1 | XP_016867340.1 | 7 - 8 | 822 | 147 | |
XM_017011852.1 | XP_016867341.1 | 7 - 8 | 972 | 147 | |
XM_017011853.1 | XP_016867342.1 | 6 - 7 | 772 | 147 | |
GenBank mRNA | AK095387.1 | - | 6 - 7 | 757 | 147 |
AK097537.1 | - | 6 - 7 | 916 | 147 | |
AY359238.1 | - | 6 - 7 | 623 | 147 | |
AY359239.1 | - | 6 - 7 | 623 | 147 | |
BC026121.1 | - | 7 - 8 | 833 | 147 | |
BC031285.1 | - | 6 - 7 | 731 | 147 |