The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172435.1 | NP_001165906.1 | 12 - 13 | 1109 | 85 |
NM_012233.2 | NP_036365.1 | 12 - 13 | 1109 | 85 | |
XM_011510823.2 | XP_011509125.1 | 12 - 13 | 1109 | 85 | |
XM_011510825.2 | XP_011509127.1 | 12 - 13 | 1109 | 85 | |
GenBank mRNA | AB449877.1 | - | 12 - 13 | 1066 | 85 |
AK022494.1 | - | 12 - 13 | 1081 | 85 | |
BC146809.1 | - | 14 - 15 | 1346 | 85 | |
D31886.1 | - | 12 - 13 | 1068 | 85 |