The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011510825.2 | XP_011509127.1 | 24 - 25 | 2955 | 127 |
GenBank mRNA | AL096752.1 | - | 5 - 6 | 1681 | 127 |
BC146809.1 | - | 26 - 27 | 3192 | 127 |