The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020410.2 | NP_065143.2 | 10 - 11 | 1425 | 77 |
GenBank mRNA | AB058728.1 | - | 10 - 11 | 1395 | 77 |
AF288687.1 | - | 10 - 11 | 1043 | 77 | |
AK056420.1 | - | 10 - 11 | 1401 | 77 | |
AK130125.1 | - | 1 - 2 | 65 | 77 | |
AK172778.1 | - | 9 - 10 | 856 | 77 |