The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014243.2 | NP_055058.2 | 17 - 18 | 2451 | 72 |
XM_011532421.2 | XP_011530723.1 | 17 - 18 | 2483 | 72 | |
XM_011532422.2 | XP_011530724.1 | 17 - 18 | 2982 | 72 | |
GenBank mRNA | AB002364.1 | - | 17 - 18 | 2403 | 72 |
BC130287.1 | - | 17 - 18 | 2448 | 72 | |
BC132735.1 | - | 17 - 18 | 2448 | 72 |