The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024551.2 | NP_078827.2 | 3 - 4 | 484 | 76 |
XM_005253789.1 | XP_005253846.1 | 3 - 4 | 606 | 76 | |
XM_006719018.2 | XP_006719081.1 | 4 - 5 | 603 | 76 | |
XM_011521024.2 | XP_011519326.1 | 4 - 5 | 614 | 76 | |
GenBank mRNA | AK127196.1 | - | 4 - 5 | 539 | 76 |
AK128511.1 | - | 3 - 4 | 538 | 76 | |
AY424280.1 | - | 2 - 3 | 363 | 76 | |
BC051858.2 | - | 3 - 4 | 484 | 76 |