The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135155.2 | NP_001128627.1 | 6 - 7 | 710 | 60 |
NM_001135156.2 | NP_001128628.1 | 6 - 7 | 724 | 60 | |
NM_001289978.1 | NP_001276907.1 | 6 - 7 | 710 | 60 | |
XM_005259289.2 | XP_005259346.1 | 6 - 7 | 710 | 60 | |
XM_005259290.3 | XP_005259347.1 | 6 - 7 | 681 | 60 | |
XM_005259292.4 | XP_005259349.1 | 6 - 7 | 647 | 60 | |
XM_006723407.2 | XP_006723470.1 | 6 - 7 | 681 | 60 | |
XM_006723408.3 | XP_006723471.1 | 6 - 7 | 649 | 60 | |
XM_011527356.1 | XP_011525658.1 | 6 - 7 | 710 | 60 | |
XM_011527357.2 | XP_011525659.1 | 6 - 7 | 650 | 60 | |
XM_011527358.2 | XP_011525660.1 | 6 - 7 | 725 | 60 | |
GenBank mRNA | AK094632.1 | - | 6 - 7 | 1037 | 60 |
AL538568.3 | - | 6 - 7 | 729 | 60 | |
BC021191.2 | - | 6 - 7 | 683 | 60 | |
BC125153.1 | - | 6 - 7 | 612 | 60 | |
GQ358728.1 | - | 6 - 7 | 602 | 60 |