The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022070.4 | NP_071353.4 | 9 - 10 | 1439 | 96 |
XM_005257570.3 | XP_005257627.1 | 10 - 11 | 1627 | 96 | |
XM_011525110.2 | XP_011523412.1 | 8 - 9 | 1326 | 96 | |
XM_011525111.2 | XP_011523413.1 | 8 - 9 | 1288 | 96 | |
XM_017024936.1 | XP_016880425.1 | 9 - 10 | 1700 | 96 | |
XM_017024937.1 | XP_016880426.1 | 9 - 10 | 3376 | 96 | |
XM_017024938.1 | XP_016880427.1 | 9 - 10 | 1432 | 96 | |
XM_017024939.1 | XP_016880428.1 | 9 - 10 | 1703 | 96 | |
GenBank mRNA | AF349752.1 | - | 10 - 11 | 1384 | 96 |
AK025740.1 | - | 2 - 3 | 506 | 96 | |
AK026070.1 | - | 2 - 3 | 279 | 96 | |
AK127708.1 | - | 9 - 10 | 1433 | 96 | |
AK314655.1 | - | 9 - 10 | 1442 | 96 | |
BC119771.1 | - | 9 - 10 | 1429 | 96 | |
BC119813.1 | - | 9 - 10 | 1430 | 96 | |
BX640831.1 | - | 9 - 10 | 1430 | 96 | |
CR627418.1 | - | 9 - 10 | 1429 | 96 |