Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- DHRS9
- Entrez Gene ID
-
10170
- Gene Name
- dehydrogenase/reductase 9
- Gene Aliases
-
3-alpha-HSD, 3ALPHA-HSD, RDH-E2, RDH-TBE, RDH15, RDHL, RDHTBE, RETSDR8, SDR9C4
- Chromosome Location
-
Chr.2: 169067035 - 169096167 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF240698.1 |
- |
4 - 5 |
743 |
83 |
View Full Product Details