The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145032.3 | NP_659469.3 | 16 - 17 | 2055 | 176 |
XM_005250205.3 | XP_005250262.1 | 17 - 18 | 1961 | 176 | |
XM_005250207.3 | XP_005250264.1 | 16 - 17 | 1862 | 176 | |
XM_005250209.2 | XP_005250266.1 | 16 - 17 | 1859 | 176 | |
XM_006715898.2 | XP_006715961.1 | 11 - 12 | 1306 | 176 | |
XM_011515929.2 | XP_011514231.1 | 17 - 18 | 1961 | 176 | |
XM_011515930.2 | XP_011514232.1 | 16 - 17 | 1925 | 176 | |
XM_017011850.1 | XP_016867339.1 | 17 - 18 | 1905 | 176 | |
XM_017011851.1 | XP_016867340.1 | 17 - 18 | 1961 | 176 | |
XM_017011852.1 | XP_016867341.1 | 17 - 18 | 2111 | 176 | |
XM_017011853.1 | XP_016867342.1 | 16 - 17 | 1911 | 176 | |
GenBank mRNA | AK095387.1 | - | 11 - 12 | 1278 | 176 |
AK097537.1 | - | 16 - 17 | 2055 | 176 | |
AL713709.1 | - | 7 - 8 | 777 | 176 | |
BC020572.2 | - | 10 - 11 | 1132 | 176 | |
BC020575.2 | - | 2 - 3 | 165 | 176 |