The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145032.3 | NP_659469.3 | 17 - 18 | 2149 | 120 |
XM_005250205.3 | XP_005250262.1 | 18 - 19 | 2055 | 120 | |
XM_005250207.3 | XP_005250264.1 | 17 - 18 | 1956 | 120 | |
XM_005250209.2 | XP_005250266.1 | 17 - 18 | 1953 | 120 | |
XM_006715898.2 | XP_006715961.1 | 12 - 13 | 1400 | 120 | |
XM_011515929.2 | XP_011514231.1 | 18 - 19 | 2055 | 120 | |
XM_011515930.2 | XP_011514232.1 | 17 - 18 | 2019 | 120 | |
XM_017011850.1 | XP_016867339.1 | 18 - 19 | 1999 | 120 | |
XM_017011851.1 | XP_016867340.1 | 18 - 19 | 2055 | 120 | |
XM_017011852.1 | XP_016867341.1 | 18 - 19 | 2205 | 120 | |
XM_017011853.1 | XP_016867342.1 | 17 - 18 | 2005 | 120 | |
GenBank mRNA | AK095387.1 | - | 12 - 13 | 1372 | 120 |
AK097537.1 | - | 17 - 18 | 2149 | 120 | |
AL713709.1 | - | 8 - 9 | 871 | 120 | |
BC020572.2 | - | 11 - 12 | 1226 | 120 | |
BC020575.2 | - | 3 - 4 | 259 | 120 |