The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111038.1 | NP_001104508.1 | 2 - 3 | 254 | 151 |
NM_001287150.1 | NP_001274079.1 | 2 - 3 | 254 | 151 | |
NM_145032.3 | NP_659469.3 | 2 - 3 | 431 | 151 | |
XM_005250209.2 | XP_005250266.1 | 2 - 3 | 235 | 151 | |
GenBank mRNA | AK097537.1 | - | 2 - 3 | 431 | 151 |
AY359238.1 | - | 2 - 3 | 138 | 151 | |
AY359239.1 | - | 2 - 3 | 138 | 151 | |
BC031285.1 | - | 2 - 3 | 246 | 151 |