The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324266.1 | NP_001311195.1 | 5 - 6 | 1042 | 155 |
NM_024576.4 | NP_078852.3 | 5 - 6 | 1039 | 155 | |
XM_011536117.2 | XP_011534419.1 | 5 - 6 | 975 | 155 | |
XM_011536118.2 | XP_011534420.1 | 5 - 6 | 824 | 155 | |
XM_017011303.1 | XP_016866792.1 | 5 - 6 | 604 | 155 | |
XM_017011304.1 | XP_016866793.1 | 5 - 6 | 601 | 155 | |
XM_017011305.1 | XP_016866794.1 | 5 - 6 | 497 | 155 | |
XM_017011306.1 | XP_016866795.1 | 5 - 6 | 1527 | 155 | |
GenBank mRNA | AK024732.1 | - | 2 - 3 | 102 | 155 |
BC018688.1 | - | 5 - 6 | 674 | 155 | |
BC110999.1 | - | 5 - 6 | 678 | 155 | |
BX420584.2 | - | 5 - 6 | 708 | 155 |