Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RALGAPA1
- Entrez Gene ID
-
253959
- Gene Name
- Ral GTPase activating protein catalytic alpha subunit 1
- Gene Aliases
-
GARNL1, GRIPE, RalGAPalpha1, TULIP1, p240
- Chromosome Location
-
Chr.14: 35538352 - 35809304 on Build GRCh38
- Gene Symbol
- RALGAPA1P1
- Entrez Gene ID
-
26134
- Gene Name
- Ral GTPase activating protein catalytic alpha subunit 1 pseudogene 1
- Gene Aliases
-
GARNL2, GARNL2P, RALGAPA1P, bA235C23.1
- Chromosome Location
-
Chr.9: 105519824 - 105527319 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_104269.1 |
|
1 - 1 |
2016 |
128 |
View Full Product Details