Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNA2
Entrez Gene ID
1763
Gene Name
DNA replication helicase/nuclease 2
Gene Aliases
DNA2L, hDNA2
Chromosome Location
Chr.10: 68414064 - 68472521 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080449.2 NP_001073918.2 1 - 2 182 88
NR_102264.1 2 - 3 162 88
XM_006717680.3 XP_006717743.1 2 - 3 562 88
GenBank mRNA AK307939.1 - 1 - 2 149 88
AK308333.1 - 1 - 2 182 88
BC063664.1 - 1 - 2 95 88
BC111740.1 - 2 - 3 162 88
D42046.1 - 1 - 2 124 88
DC404138.1 - 1 - 2 183 89

View Full Product Details