The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024496.3 | NP_078772.1 | 1 - 1 | 4020 | 78 |
GenBank mRNA | AB058768.1 | - | 2 - 2 | 3506 | 78 |
AL832320.1 | - | 1 - 1 | 2112 | 78 | |
BC108292.1 | - | 1 - 1 | 4010 | 78 | |
BC152431.1 | - | 2 - 2 | 3506 | 78 | |
CB852902.1 | - | 1 - 1 | 78 |