Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSPAN15
Entrez Gene ID
23555
Gene Name
tetraspanin 15
Gene Aliases
2700063A19Rik, NET-7, NET7, TM4SF15
Chromosome Location
Chr.10: 69451464 - 69549508 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012339.3 NP_036471.1 6 - 7 744 67
XM_005269666.4 XP_005269723.1 4 - 5 1204 67
XM_005269667.3 XP_005269724.1 4 - 5 487 67
XM_011539562.2 XP_011537864.1 4 - 5 447 67
XM_011539563.2 XP_011537865.1 4 - 5 658 67
GenBank mRNA AF120266.1 - 6 - 7 735 67
BC003157.1 - 6 - 7 738 67
BC004161.2 - 6 - 7 738 67

View Full Product Details