The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318766.1 | NP_001305695.1 | 12 - 13 | 1406 | 73 |
NM_033388.1 | NP_203746.1 | 12 - 13 | 1286 | 73 | |
XM_005274376.4 | XP_005274433.1 | 12 - 13 | 1285 | 73 | |
XM_006718732.2 | XP_006718795.1 | 11 - 12 | 1222 | 73 | |
XM_006718734.2 | XP_006718797.1 | 4 - 5 | 2465 | 73 | |
XM_011545332.1 | XP_011543634.1 | 11 - 12 | 1171 | 73 | |
XM_011545333.1 | XP_011543635.1 | 12 - 13 | 1734 | 73 | |
XM_011545334.1 | XP_011543636.1 | 12 - 13 | 1147 | 73 | |
GenBank mRNA | AK090597.1 | - | 12 - 13 | 1397 | 73 |
AK091151.1 | - | 11 - 12 | 2874 | 73 | |
AK093017.1 | - | 12 - 13 | 1259 | 73 | |
AL832974.1 | - | 4 - 5 | 897 | 73 | |
BC036713.2 | - | 10 - 11 | 1015 | 73 | |
BC137489.1 | - | 12 - 13 | 1384 | 73 | |
BC137490.1 | - | 12 - 13 | 1384 | 73 | |
BC142718.1 | - | 10 - 11 | 2982 | 73 | |
BC146660.1 | - | 10 - 11 | 2982 | 73 |