Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATG16L2
Entrez Gene ID
89849
Gene Name
autophagy related 16 like 2
Gene Aliases
ATG16B, WDR80
Chromosome Location
Chr.11: 72814406 - 72843740 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318766.1 NP_001305695.1 15 - 16 1782 67
NM_033388.1 NP_203746.1 15 - 16 1662 67
XM_005274376.4 XP_005274433.1 15 - 16 1661 67
XM_006718732.2 XP_006718795.1 14 - 15 1598 67
XM_006718734.2 XP_006718797.1 7 - 8 2841 67
XM_011545332.1 XP_011543634.1 14 - 15 1547 67
XM_011545333.1 XP_011543635.1 15 - 16 2110 67
XM_011545334.1 XP_011543636.1 15 - 16 1523 67
GenBank mRNA AK090597.1 - 15 - 16 1773 67
AK091151.1 - 14 - 15 3250 67
AK093017.1 - 15 - 16 1635 67
AL832974.1 - 7 - 8 1273 67
BC036713.2 - 13 - 14 1391 67
BC137489.1 - 15 - 16 1760 67
BC137490.1 - 15 - 16 1760 67
BC142718.1 - 13 - 14 3358 67
BC146660.1 - 13 - 14 3358 67
BQ929430.1 - 4 - 5 519 67

View Full Product Details