The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018060.3 | NP_060530.3 | 20 - 21 | 2665 | 104 |
GenBank mRNA | AK001188.1 | - | 18 - 19 | 2140 | 104 |
AK001726.1 | - | 14 - 15 | 1653 | 104 | |
AK022665.1 | - | 10 - 11 | 1232 | 104 | |
AK291432.1 | - | 20 - 21 | 2474 | 104 | |
AY267462.1 | - | 20 - 21 | 2676 | 104 | |
BC010218.1 | - | 16 - 17 | 1845 | 104 | |
BC040376.1 | - | 20 - 21 | 2398 | 104 | |
BC047880.1 | - | 19 - 20 | 2192 | 104 | |
BC137438.1 | - | 20 - 21 | 2635 | 104 | |
D28500.1 | - | 20 - 21 | 2506 | 104 |