The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202521.1 | NP_001189450.1 | 8 - 9 | 1240 | 102 |
NM_001202522.1 | NP_001189451.1 | 8 - 9 | 1240 | 102 | |
NM_001202523.1 | NP_001189452.1 | 9 - 10 | 1492 | 102 | |
NM_001297652.1 | NP_001284581.1 | 9 - 10 | 1611 | 102 | |
NM_001297653.1 | NP_001284582.1 | 9 - 10 | 1413 | 102 | |
NM_001297654.1 | NP_001284583.1 | 9 - 10 | 1413 | 102 | |
NM_001954.4 | NP_001945.3 | 11 - 12 | 1531 | 102 | |
NM_013993.2 | NP_054699.2 | 10 - 11 | 1457 | 102 | |
NM_013994.2 | NP_054700.2 | 8 - 9 | 1240 | 102 | |
XM_006715185.2 | XP_006715248.1 | 9 - 10 | 1404 | 102 | |
XM_011514882.2 | XP_011513184.1 | 9 - 10 | 1404 | 102 | |
XM_011514883.1 | XP_011513185.1 | 11 - 12 | 1519 | 102 | |
XM_011514884.1 | XP_011513186.1 | 11 - 12 | 1595 | 102 | |
XM_011514885.1 | XP_011513187.1 | 9 - 10 | 1442 | 102 | |
XM_011514886.1 | XP_011513188.1 | 9 - 10 | 1307 | 102 | |
XM_011514887.1 | XP_011513189.1 | 9 - 10 | 1457 | 102 | |
XM_011514888.2 | XP_011513190.1 | 9 - 10 | 1404 | 102 | |
XM_017011268.1 | XP_016866757.1 | 9 - 10 | 1274 | 102 | |
XM_017011269.1 | XP_016866758.1 | 9 - 10 | 1457 | 102 | |
GenBank mRNA | AB210021.1 | - | 10 - 11 | 1352 | 102 |
AK130776.1 | - | 4 - 5 | 586 | 102 | |
AK291621.1 | - | 9 - 10 | 1306 | 102 | |
AK294793.1 | - | 9 - 10 | 1478 | 102 | |
BC008716.2 | - | 11 - 12 | 1531 | 102 | |
BC013400.2 | - | 11 - 12 | 1531 | 102 | |
BC070070.1 | - | 9 - 10 | 1302 | 102 | |
L11315.1 | - | 10 - 11 | 1339 | 102 | |
L20817.1 | - | 9 - 10 | 1390 | 102 | |
L57508.1 | - | 9 - 10 | 1390 | 102 | |
X74979.1 | - | 8 - 9 | 1243 | 102 | |
Z29093.1 | - | 8 - 9 | 1534 | 102 |