The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077365.1 | NP_001070833.1 | 19 - 20 | 2207 | 67 |
NM_001077366.1 | NP_001070834.1 | 18 - 19 | 2055 | 67 | |
NM_001136113.1 | NP_001129585.1 | 19 - 20 | 2447 | 67 | |
NM_001136114.1 | NP_001129586.1 | 17 - 18 | 1948 | 67 | |
NM_007171.3 | NP_009102.3 | 19 - 20 | 2273 | 67 | |
XM_005272156.1 | XP_005272213.1 | 19 - 20 | 2399 | 67 | |
XM_005272158.1 | XP_005272215.1 | 18 - 19 | 2128 | 67 | |
XM_005272159.1 | XP_005272216.1 | 17 - 18 | 2018 | 67 | |
XM_005272162.2 | XP_005272219.1 | 18 - 19 | 2449 | 67 | |
XM_006716932.1 | XP_006716995.1 | 18 - 19 | 2051 | 67 | |
XM_011518140.1 | XP_011516442.1 | 18 - 19 | 2126 | 67 | |
XM_011518141.1 | XP_011516443.1 | 18 - 19 | 2060 | 67 | |
XM_011518142.1 | XP_011516444.1 | 17 - 18 | 1928 | 67 | |
XM_011518143.1 | XP_011516445.1 | 17 - 18 | 1958 | 67 | |
XM_011518145.1 | XP_011516447.1 | 16 - 17 | 1794 | 67 | |
XM_017014203.1 | XP_016869692.1 | 19 - 20 | 2721 | 67 | |
XM_017014204.1 | XP_016869693.1 | 18 - 19 | 1995 | 67 | |
XM_017014205.1 | XP_016869694.1 | 19 - 20 | 2200 | 67 | |
GenBank mRNA | AB209430.1 | - | 13 - 14 | 3903 | 67 |
AK000475.1 | - | 8 - 9 | 862 | 67 | |
AK074874.1 | - | 19 - 20 | 2165 | 67 | |
AK074888.1 | - | 18 - 19 | 2039 | 67 | |
AK094558.1 | - | 9 - 10 | 2555 | 67 | |
AK293529.1 | - | 17 - 18 | 1860 | 67 | |
AK295458.1 | - | 12 - 13 | 1372 | 67 | |
AK295561.1 | - | 17 - 18 | 1952 | 67 | |
AK299646.1 | - | 16 - 17 | 1759 | 67 | |
AK300393.1 | - | 18 - 19 | 2075 | 67 | |
AK315218.1 | - | 18 - 19 | 2410 | 67 | |
BC022877.2 | - | 19 - 20 | 2101 | 67 | |
BC065268.1 | - | 19 - 20 | 2392 | 67 | |
BF984005.1 | - | - | 224 | 67 |