Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VARS2
Entrez Gene ID
57176
Gene Name
valyl-tRNA synthetase 2, mitochondrial
Gene Aliases
COXPD20, VALRS, VARS2L, VARSL
Chromosome Location
Chr.6: 30914208 - 30926459 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167733.2 NP_001161205.1 11 - 12 1105 70
NM_001167734.1 NP_001161206.1 12 - 13 1340 70
NM_020442.5 NP_065175.4 12 - 13 1333 70
GenBank mRNA AB067472.1 - 11 - 12 1801 70
AK000511.1 - 11 - 12 1010 70
AK125069.1 - 1 - 2 1089 70
AK293780.1 - 11 - 12 1094 70
AK303415.1 - 12 - 13 1322 70
AK304807.1 - 12 - 13 1340 70
AK315733.1 - 1 - 2 91 70
BC009355.2 - 2 - 3 181 70
BC112054.1 - 11 - 12 1195 70
BC113605.1 - 11 - 12 1195 70
BC143534.1 - 11 - 12 1194 70
BC143535.1 - 11 - 12 1194 70

View Full Product Details