Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VARS2
Entrez Gene ID
57176
Gene Name
valyl-tRNA synthetase 2, mitochondrial
Gene Aliases
COXPD20, VALRS, VARS2L, VARSL
Chromosome Location
Chr.6: 30914208 - 30926459 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167733.2 NP_001161205.1 2 - 3 226 82
NM_001167734.1 NP_001161206.1 3 - 4 461 82
NM_020442.5 NP_065175.4 3 - 4 454 82
GenBank mRNA AB067472.1 - 2 - 3 922 82
AK000511.1 - 3 - 4 341 82
AK293780.1 - 2 - 3 215 82
AK294451.1 - 3 - 4 398 82
AK303415.1 - 3 - 4 443 82
AK304807.1 - 3 - 4 461 82
BC112054.1 - 2 - 3 316 82
BC113605.1 - 2 - 3 316 82
BC143534.1 - 2 - 3 315 82
BC143535.1 - 2 - 3 315 82
BC143536.1 - 2 - 3 315 82
DC307585.1 - 3 - 4 460 82

View Full Product Details