The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032854.3 | NP_116243.2 | 8 - 9 | 1066 | 98 |
XM_005258047.3 | XP_005258104.1 | 9 - 10 | 1348 | 98 | |
XM_005258048.4 | XP_005258105.2 | 9 - 10 | 1285 | 98 | |
XM_005258056.4 | XP_005258113.1 | 4 - 5 | 1165 | 98 | |
XM_011525385.2 | XP_011523687.1 | 9 - 10 | 1285 | 98 | |
XM_011525386.2 | XP_011523688.1 | 8 - 9 | 1180 | 98 | |
XM_011525387.2 | XP_011523689.1 | 8 - 9 | 1162 | 98 | |
XM_011525390.2 | XP_011523692.1 | 5 - 6 | 1028 | 98 | |
XM_011525391.2 | XP_011523693.1 | 5 - 6 | 1006 | 98 | |
XM_017025235.1 | XP_016880724.1 | 8 - 9 | 1229 | 98 | |
GenBank mRNA | AF193039.1 | - | 3 - 4 | 267 | 98 |
AF447884.1 | - | 5 - 6 | 1050 | 98 | |
AF447885.1 | - | 3 - 4 | 736 | 98 | |
AK027777.1 | - | 3 - 4 | 738 | 98 | |
AK094683.1 | - | 4 - 5 | 1888 | 98 | |
AK096431.1 | - | 8 - 9 | 971 | 98 | |
AL833952.1 | - | 6 - 7 | 2457 | 98 | |
AY545070.1 | - | 3 - 4 | 506 | 98 | |
BC048209.1 | - | 5 - 6 | 480 | 98 | |
BC064514.1 | - | 5 - 6 | 480 | 98 |