Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CORO6
Entrez Gene ID
84940
Gene Name
coronin 6
Gene Aliases
-
Chromosome Location
Chr.17: 29614756 - 29622912 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011525389.2 XP_011523691.1 5 - 6 850 80

View Full Product Details