Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CORO6
Entrez Gene ID
84940
Gene Name
coronin 6
Gene Aliases
-
Chromosome Location
Chr.17: 29614756 - 29622912 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032854.3 NP_116243.2 5 - 6 753 69
XM_005258047.3 XP_005258104.1 6 - 7 1035 69
XM_005258048.4 XP_005258105.2 6 - 7 972 69
XM_011525385.2 XP_011523687.1 6 - 7 972 69
XM_011525387.2 XP_011523689.1 5 - 6 849 69
XM_011525388.2 XP_011523690.1 6 - 7 972 69
XM_017025235.1 XP_016880724.1 5 - 6 916 69
GenBank mRNA AK096431.1 - 5 - 6 658 69

View Full Product Details