The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077366.1 | NP_001070834.1 | 1 - 2 | 173 | 107 |
XM_005272158.1 | XP_005272215.1 | 1 - 2 | 180 | 107 | |
XM_011518142.1 | XP_011516444.1 | 1 - 2 | 127 | 107 | |
XM_017014204.1 | XP_016869693.1 | 1 - 2 | 108 | 107 | |
GenBank mRNA | AK074888.1 | - | 1 - 2 | 157 | 107 |